Assay Details
| Gene Symbol: Sult1c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031732.2 | NP_113920.2 | 1 - 2 | 89 | 110 | |
| GenBank mRNA | BC088125.1 | - | 1 - 2 | 89 | 7324981 | 110 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC088125.1,NM_031732.2 transcripts.
Set Membership: |
|

Sult1c3
Genomic Map