Assay Details
| Gene Symbol: Sult1c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031732.2 | NP_113920.2 | 2 - 3 | 280 | 111 | |
| GenBank mRNA | BC088125.1 | - | 2 - 3 | 280 | 7324981 | 111 |
| FQ219411.1 | - | 2 - 3 | 310 | 111 | ||
| L22339.1 | - | 1 - 2 | 190 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sult1c3
Genomic Map