Assay Details
| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037660.1 | NP_001032749.1 | 5 - 6 | 514 | 105 | |
| GenBank mRNA | AJ628424.1 | - | 5 - 6 | 514 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csf2ra
Genomic Map