Assay Details
| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032617.2 | NP_116006.1 | 2 - 3 | 280 | 120 | |
| GenBank mRNA | AF286534.1 | - | 2 - 3 | 286 | 120 | |
| BC062041.1 | - | 2 - 3 | 280 | 5598168 | 120 | |
| D01046.1 | - | 2 - 3 | 266 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11b
Genomic Map