Assay Details
| Gene Symbol: Ccdc132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173511.1 | NP_001166982.1 | 21 - 22 | 2057 | 127 | |
| XM_006236037.2 | XP_006236099.1 | 21 - 22 | 2088 | 127 | ||
| XM_008762704.1 | XP_008760926.1 | 21 - 22 | 2091 | 127 | ||
| GenBank mRNA | BC091288.1 | - | 6 - 7 | 652 | 127 | |
| Gene Symbol: LOC100911994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749697.3 | XP_003749745.2 | 1 - 2 | 1315 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc132
Genomic Map