Assay Details
| Gene Symbol: Myl6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109486.1 | NP_001102956.1 | 6 - 7 | 447 | 99 | |
| Gene Symbol: Myl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765040.1 | XP_008763262.1 | 6 - 7 | 541 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myl6b
Genomic Map