Assay Details
| Gene Symbol: Ssr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017199.1 | NP_058895.1 | 2 - 3 | 344 | 133 | |
| XM_006229571.2 | XP_006229633.1 | 2 - 3 | 400 | 133 | ||
| GenBank mRNA | BC058482.1 | - | 2 - 3 | 257 | 6921620 | 133 |
| Z19087.1 | - | 2 - 3 | 344 | 133 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ssr4
Genomic Map