Assay Details
| Gene Symbol: Rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031100.1 | NP_112362.1 | 2 - 3 | 80 | 80 | |
| GenBank mRNA | BC058467.1 | - | 2 - 3 | 69 | 6918973 | 80 |
| FQ210278.1 | - | 2 - 3 | 64 | 80 | ||
| FQ218098.1 | - | 2 - 3 | 64 | 80 | ||
| FQ220555.1 | - | 2 - 3 | 63 | 80 | ||
| FQ221864.1 | - | 2 - 3 | 64 | 80 | ||
| FQ222021.1 | - | 2 - 3 | 64 | 80 | ||
| FQ229032.1 | - | 2 - 3 | 63 | 80 | ||
| FQ233952.1 | - | 2 - 3 | 64 | 80 | ||
| X87106.1 | - | 2 - 3 | 80 | 80 | ||
| Gene Symbol: RGD1563861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_574109.5 | XP_574109.1 | 1 | 65 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC058467.1,NM_031100.1,X87106.1,XM_574109.5 transcripts.
This assay is in the 5'UTR region of the BC058467.1,NM_031100.1,X87106.1,XM_574109.5 transcripts.
Set Membership: |
|

Rpl10
Genomic Map