Assay Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766411.1 | XP_008764633.1 | 7 - 8 | 1747 | 95 | |
| XM_343420.8 | XP_343421.3 | 7 - 8 | 1739 | 95 | ||
More Information
Set Membership: |
|

Fam63b
Genomic Map