Assay Details
| Gene Symbol: Dyx1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007010.1 | NP_001007011.1 | 5 - 6 | 867 | 56 | |
| XM_006243328.2 | XP_006243390.1 | 5 - 6 | 947 | 56 | ||
| XM_006243329.2 | XP_006243391.1 | 5 - 6 | 947 | 56 | ||
| XM_006243331.2 | XP_006243393.1 | 3 - 4 | 587 | 56 | ||
| XM_008766313.1 | XP_008764535.1 | 5 - 6 | 845 | 56 | ||
| XM_008766314.1 | XP_008764536.1 | 5 - 6 | 676 | 56 | ||
| GenBank mRNA | AY428506.1 | - | 5 - 6 | 867 | 56 | |
| BC085838.1 | - | 5 - 6 | 811 | 7135744 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dyx1c1
Genomic Map