Assay Details
| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169142.1 | NP_001162613.1 | 8 - 9 | 2904 | 53 | |
| XM_008758753.1 | XP_008756975.1 | 8 - 9 | 3071 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp12
Genomic Map