Assay Details
| Gene Symbol: LOC292543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025639.1 | NP_001020810.1 | 1 - 2 | 63 | 111 | |
| XM_008758754.1 | XP_008756976.1 | 1 - 2 | 219 | 111 | ||
| GenBank mRNA | BC098062.1 | - | 1 - 2 | 63 | 7458331 | 111 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC098062.1, NM_001025639.1 transcripts.
Set Membership: |
|

LOC292543
Genomic Map