Assay Details
| Gene Symbol: LOC292543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025639.1 | NP_001020810.1 | 5 - 6 | 913 | 79 | |
| XM_006228022.2 | XP_006228084.1 | 5 - 6 | 1005 | 79 | ||
| XM_008758754.1 | XP_008756976.1 | 5 - 6 | 1078 | 79 | ||
| GenBank mRNA | BC098062.1 | - | 5 - 6 | 913 | 7458331 | 79 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC292543
Genomic Map