Assay Details
| Gene Symbol: Ube2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031001.2 | NP_112263.1 | 1 | 1203 | 68 | |
| GenBank mRNA | BC078808.1 | - | 1 | 1203 | 7113229 | 68 |
| U56407.1 | - | 1 | 1094 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ube2d4
Genomic Map