Assay Details
| Gene Symbol: Tmed5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007619.1 | NP_001007620.1 | 2 - 3 | 364 | 87 | |
| GenBank mRNA | AY539871.1 | - | 276 | 87 | ||
| BC079452.1 | - | 2 - 3 | 364 | 7121068 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmed5
Genomic Map