Assay Details
| Gene Symbol: Rpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031099.1 | NP_112361.1 | 3 - 4 | 313 | 129 | |
| XM_006250585.2 | XP_006250647.1 | 3 - 4 | 294 | 129 | ||
| GenBank mRNA | BC060561.1 | - | 3 - 4 | 260 | 6918805 | 129 |
| FQ212710.1 | - | 3 - 4 | 270 | 129 | ||
| FQ214281.1 | - | 3 - 4 | 261 | 129 | ||
| FQ214493.1 | - | 3 - 4 | 261 | 129 | ||
| FQ215029.1 | - | 3 - 4 | 262 | 129 | ||
| FQ215486.1 | - | 2 - 3 | 134 | 129 | ||
| FQ219244.1 | - | 3 - 4 | 259 | 129 | ||
| FQ220068.1 | - | 3 - 4 | 261 | 129 | ||
| FQ220161.1 | - | 3 - 4 | 261 | 129 | ||
| FQ220170.1 | - | 3 - 4 | 261 | 129 | ||
| FQ220205.1 | - | 3 - 4 | 261 | 129 | ||
| FQ220219.1 | - | 3 - 4 | 261 | 129 | ||
| FQ227439.1 | - | 3 - 4 | 261 | 129 | ||
| FQ228473.1 | - | 3 - 4 | 261 | 129 | ||
| FQ229451.1 | - | 3 - 4 | 262 | 129 | ||
| FQ229816.1 | - | 3 - 4 | 259 | 129 | ||
| FQ229979.1 | - | 3 - 4 | 261 | 129 | ||
| FQ230017.1 | - | 3 - 4 | 259 | 129 | ||
| FQ230061.1 | - | 3 - 4 | 264 | 129 | ||
| FQ230102.1 | - | 3 - 4 | 261 | 129 | ||
| FQ230128.1 | - | 3 - 4 | 289 | 129 | ||
| FQ230161.1 | - | 3 - 4 | 287 | 129 | ||
| FQ230794.1 | - | 3 - 4 | 264 | 129 | ||
| FQ231729.1 | - | 3 - 4 | 259 | 129 | ||
| FQ232540.1 | - | 3 - 4 | 261 | 129 | ||
| FQ233314.1 | - | 3 - 4 | 286 | 129 | ||
| FQ234171.1 | - | 3 - 4 | 291 | 129 | ||
| M17419.1 | - | 3 - 4 | 246 | 129 | ||
| X06148.1 | - | 3 - 4 | 313 | 129 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl5
Genomic Map