Assay Details
| Gene Symbol: Ephx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105994.1 | NP_001099464.1 | 1 - 2 | 191 | 103 | |
| XM_006250541.1 | XP_006250603.1 | 1 - 2 | 244 | 103 | ||
| GenBank mRNA | FQ212126.1 | - | 1 - 2 | 215 | 103 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001105994.1 transcripts.
Set Membership: |
|

Ephx4
Genomic Map