Assay Details
| Gene Symbol: Ephx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105994.1 | NP_001099464.1 | 3 - 4 | 429 | 84 | |
| XM_006250541.1 | XP_006250603.1 | 3 - 4 | 482 | 84 | ||
| XM_008769927.1 | XP_008768149.1 | 3 - 4 | 1341 | 84 | ||
| GenBank mRNA | FQ212126.1 | - | 3 - 4 | 453 | 84 | |
| FQ213206.1 | - | 3 - 4 | 355 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephx4
Genomic Map