Assay Details
| Gene Symbol: Zfp57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213565.4 | NP_998730.2 | 4 - 5 | 433 | 124 | |
| XM_008772694.1 | XP_008770916.1 | 3 - 4 | 606 | 124 | ||
| GenBank mRNA | BC127468.1 | - | 2 - 3 | 251 | 8368087 | 124 |
| CA511208.1 | - | 4 - 5 | 434 | 124 | ||
| DY312591.1 | - | 4 - 5 | 485 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp57
Genomic Map