Assay Details
| Gene Symbol: Large | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108439.1 | NP_001101909.1 | 1 - 2 | 384 | 64 | |
| XM_006255140.2 | XP_006255202.1 | 1 - 2 | 1035 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108439.1, XM_006255140.2 transcripts.
Set Membership: |
|

Large
Genomic Map