Assay Details
| Gene Symbol: Large | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108439.1 | NP_001101909.1 | 5 - 6 | 1085 | 107 | |
| XM_006255140.2 | XP_006255202.1 | 5 - 6 | 1736 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Large
Genomic Map