Assay Details
| Gene Symbol: Tuba1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044270.2 | NP_001037735.1 | 1 - 2 | 98 | 60 | |
| GenBank mRNA | BC060572.1 | - | 1 - 2 | 102 | 6920989 | 60 |
| BC076379.1 | - | 1 - 2 | 84 | 7106502 | 60 | |
| FQ212426.1 | - | 1 - 2 | 93 | 60 | ||
| FQ212740.1 | - | 1 - 2 | 93 | 60 | ||
| FQ213098.1 | - | 1 - 2 | 98 | 60 | ||
| FQ213226.1 | - | 1 - 2 | 93 | 60 | ||
| FQ213439.1 | - | 1 - 2 | 93 | 60 | ||
| FQ213947.1 | - | 1 - 2 | 93 | 60 | ||
| FQ214075.1 | - | 1 - 2 | 96 | 60 | ||
| FQ214301.1 | - | 1 - 2 | 96 | 60 | ||
| FQ223081.1 | - | 1 - 2 | 96 | 60 | ||
| FQ225339.1 | - | 1 - 2 | 93 | 60 | ||
| FQ225425.1 | - | 1 - 2 | 93 | 60 | ||
| FQ225650.1 | - | 1 - 2 | 93 | 60 | ||
| FQ231550.1 | - | 1 - 2 | 93 | 60 | ||
| FQ232527.1 | - | 1 - 2 | 97 | 60 | ||
| FQ233000.1 | - | 1 - 2 | 95 | 60 | ||
| FQ233566.1 | - | 1 - 2 | 96 | 60 | ||
| FQ234135.1 | - | 1 - 2 | 93 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC060572.1,BC076379.1,NM_001044270.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tuba1b
Genomic Map