Assay Details
| Gene Symbol: Itgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013171.1 | NP_037303.1 | 7 - 8 | 1162 | 64 | |
| XM_006242383.2 | XP_006242445.1 | 8 - 9 | 1258 | 64 | ||
| GenBank mRNA | AF003598.1 | - | 5 - 6 | 661 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb7
Genomic Map