Assay Details
| Gene Symbol: Igfbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013104.3 | NP_037236.1 | 2 - 3 | 442 | 136 | |
| GenBank mRNA | BC099742.1 | - | 2 - 3 | 454 | 7937967 | 136 |
| FQ220221.1 | - | 2 - 3 | 512 | 136 | ||
| FQ220720.1 | - | 2 - 3 | 513 | 136 | ||
| FQ220764.1 | - | 2 - 3 | 515 | 136 | ||
| FQ230155.1 | - | 2 - 3 | 518 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igfbp6
Genomic Map