Assay Details
| Gene Symbol: Vps72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107695.1 | NP_001101165.1 | 1 - 2 | 180 | 97 | |
| XM_008761302.1 | XP_008759524.1 | 1 - 2 | 212 | 97 | ||
| GenBank mRNA | BC161999.1 | - | 1 - 2 | 168 | 9036078 | 97 |
More Information
Updated Mapping Information
Set Membership: |
|

Vps72
Genomic Map