Assay Details
| Gene Symbol: Vps72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107695.1 | NP_001101165.1 | 3 - 4 | 450 | 75 | |
| XM_008761302.1 | XP_008759524.1 | 3 - 4 | 482 | 75 | ||
| GenBank mRNA | BC161999.1 | - | 3 - 4 | 438 | 9036078 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|

Vps72
Genomic Map