Assay Details
| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233102.2 | XP_006233164.1 | 4 - 5 | 800 | 117 | |
| XM_006233103.2 | XP_006233165.1 | 5 - 6 | 1759 | 117 | ||
| XM_008761434.1 | XP_008759656.1 | 6 - 7 | 1358 | 117 | ||
| XM_008761435.1 | XP_008759657.1 | 4 - 5 | 824 | 117 | ||
More Information
Set Membership: |
|

Fam19a3
Genomic Map