Assay Details
| Gene Symbol: Cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182672.2 | NP_872613.1 | 1 - 2 | 246 | 95 | |
| GenBank mRNA | AY305858.1 | - | 1 - 2 | 247 | 95 | |
| BC086378.1 | - | 1 - 2 | 213 | 7302014 | 95 | |
| FQ215409.1 | - | 1 - 2 | 267 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cbr4
Genomic Map