Assay Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762397.1 | XP_008760619.1 | 4 - 5 | 648 | 82 | |
| XM_008762398.1 | XP_008760620.1 | 3 - 4 | 2221 | 82 | ||
| XM_008762399.1 | XP_008760621.1 | 4 - 5 | 672 | 82 | ||
| XM_342547.7 | XP_342548.5 | 4 - 5 | 699 | 82 | ||
More Information
Set Membership: |
|

Nol4l
Genomic Map