Assay Details
| Gene Symbol: Ttll9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014051.1 | NP_001014073.1 | 8 - 9 | 720 | 61 | |
| XM_006235276.2 | XP_006235338.1 | 8 - 9 | 1491 | 61 | ||
| XM_008762331.1 | XP_008760553.1 | 9 - 10 | 775 | 61 | ||
| XM_008762332.1 | XP_008760554.1 | 9 - 10 | 775 | 61 | ||
| XM_008762333.1 | XP_008760555.1 | 8 - 9 | 589 | 61 | ||
| GenBank mRNA | BC082105.1 | - | 8 - 9 | 720 | 7134735 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Ttll9
Genomic Map