Assay Details
| Gene Symbol: Cox4i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053472.1 | NP_445924.1 | 2 - 3 | 100 | 127 | |
| XM_006235280.2 | XP_006235342.1 | 2 - 3 | 455 | 127 | ||
| GenBank mRNA | AF255347.1 | - | 2 - 3 | 100 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cox4i2
Genomic Map