Assay Details
| Gene Symbol: LOC690309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762447.1 | XP_008760669.1 | 15 - 16 | 1900 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690309| Gene Symbol: LOC690309 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762447.1 | XP_008760669.1 | 15 - 16 | 1900 | 80 | |
Set Membership: |
|