Assay Details
| Gene Symbol: Bpifb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107791.1 | NP_001101261.1 | 3 - 4 | 219 | 79 | |
| XM_006235317.1 | XP_006235379.1 | 12 - 13 | 1179 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bpifb6
Genomic Map