Assay Details
| Gene Symbol: Ces2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100477.1 | NP_001093947.1 | 1 - 2 | 78 | 70 | |
| GenBank mRNA | D50580.1 | - | 1 - 2 | 70 | 70 | |
| FQ218265.1 | - | 1 - 2 | 118 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ces2e
Genomic Map