Assay Details
| Gene Symbol: Ces2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100477.1 | NP_001093947.1 | 3 - 4 | 416 | 59 | |
| GenBank mRNA | D50580.1 | - | 3 - 4 | 408 | 59 | |
| FQ218265.1 | - | 3 - 4 | 456 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ces2e
Genomic Map