Assay Details
| Gene Symbol: Ces2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100477.1 | NP_001093947.1 | 6 - 7 | 916 | 84 | |
| GenBank mRNA | D50580.1 | - | 6 - 7 | 908 | 84 | |
| FQ218265.1 | - | 6 - 7 | 956 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ces2e
Genomic Map