Assay Details
| Gene Symbol: Cmtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106164.1 | NP_001099634.1 | 3 - 4 | 464 | 77 | |
| XM_006255049.2 | XP_006255111.1 | 4 - 5 | 547 | 77 | ||
| XM_008772241.1 | XP_008770463.1 | 4 - 5 | 551 | 77 | ||
| XM_008772242.1 | XP_008770464.1 | 4 - 5 | 500 | 77 | ||
| GenBank mRNA | BC169107.1 | - | 4 - 5 | 556 | 7130835 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm3
Genomic Map