Assay Details
| Gene Symbol: Ctgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022266.2 | NP_071602.1 | 3 - 4 | 757 | 89 | |
| GenBank mRNA | AB023068.1 | - | 3 - 4 | 745 | 89 | |
| AF120275.1 | - | 3 - 4 | 757 | 89 | ||
| AJ236872.1 | - | 1 - 2 | 256 | 89 | ||
| BC072503.1 | - | 3 - 4 | 747 | 7098774 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctgf
Genomic Map