Assay Details
| Gene Symbol: Fam3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106027.1 | NP_001099497.1 | 7 - 8 | 663 | 116 | |
| XM_006251752.2 | XP_006251814.1 | 7 - 8 | 1368 | 116 | ||
| XM_006251753.2 | XP_006251815.1 | 7 - 8 | 688 | 116 | ||
| XM_008770502.1 | XP_008768724.1 | 2 - 3 | 1621 | 116 | ||
| XM_008770503.1 | XP_008768725.1 | 5 - 6 | 1074 | 116 | ||
| XM_008770504.1 | XP_008768726.1 | 6 - 7 | 792 | 116 | ||
| XM_008770505.1 | XP_008768727.1 | 7 - 8 | 2296 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam3d
Genomic Map