Assay Details
| Gene Symbol: Rpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139114.3 | NP_620814.1 | 1 - 2 | 68 | 126 | |
| GenBank mRNA | AA687059.1 | - | 1 - 2 | 74 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_139114.3 transcripts.
Set Membership: |
|

Rpl15
Genomic Map