Assay Details
| Gene Symbol: Flnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107288.1 | NP_001100758.1 | 29 - 30 | 5234 | 137 | |
| Gene Symbol: LOC100911261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770602.1 | XP_008768824.1 | 1606 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Flnb
Genomic Map