Assay Details
| Gene Symbol: LOC100911261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751445.3 | XP_003751493.2 | 2193 | 92 | ||
| XM_008770601.1 | XP_008768823.1 | 2226 | 92 | |||
| XM_008770602.1 | XP_008768824.1 | 2154 | 92 | |||
| Gene Symbol: Flnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107288.1 | NP_001100758.1 | 33 - 34 | 5782 | 92 | |
| XM_006251780.2 | XP_006251842.1 | 34 - 35 | 5915 | 92 | ||
| XM_006251781.2 | XP_006251843.1 | 34 - 35 | 5882 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911261
Genomic Map