Assay Details
| Gene Symbol: LOC100911261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751445.3 | XP_003751493.2 | 2828 | 70 | ||
| XM_008770601.1 | XP_008768823.1 | 2861 | 70 | |||
| XM_008770602.1 | XP_008768824.1 | 2789 | 70 | |||
| Gene Symbol: Flnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107288.1 | NP_001100758.1 | 37 - 38 | 6417 | 70 | |
| XM_006251780.2 | XP_006251842.1 | 38 - 39 | 6550 | 70 | ||
| XM_006251781.2 | XP_006251843.1 | 38 - 39 | 6517 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911261
Genomic Map