Assay Details
| Gene Symbol: Flnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107288.1 | NP_001100758.1 | 40 - 41 | 6938 | 86 | |
| XM_006251780.2 | XP_006251842.1 | 41 - 42 | 7071 | 86 | ||
| XM_006251781.2 | XP_006251843.1 | 41 - 42 | 7038 | 86 | ||
| Gene Symbol: LOC100911261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751445.3 | XP_003751493.2 | 3349 | 86 | ||
| XM_008770601.1 | XP_008768823.1 | 3382 | 86 | |||
| XM_008770602.1 | XP_008768824.1 | 3310 | 86 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Flnb
Genomic Map