Assay Details
| Gene Symbol: Pou6f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101002.1 | NP_001094472.1 | 2 - 3 | 438 | 57 | |
| GenBank mRNA | BC128774.1 | - | 2 - 3 | 438 | 7132745 | 57 |
More Information
Updated Mapping Information
Set Membership: |
|

Pou6f2
Genomic Map