Assay Details
| Gene Symbol: Mtpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107359.1 | NP_001100829.1 | 1 - 2 | 177 | 84 | |
| XM_006254071.2 | XP_006254133.1 | 1 - 2 | 261 | 84 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107359.1 transcripts.
Set Membership: |
|

Mtpap
Genomic Map