Assay Details
| Gene Symbol: Oaz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109899.2 | NP_001103369.1 | 3 - 4 | 517 | 85 | |
| NM_001301297.1 | NP_001288226.1 | 3 - 4 | 514 | 85 | ||
| GenBank mRNA | CB586487.1 | - | 3 - 4 | 516 | 85 | |
| FQ211793.1 | - | 3 - 4 | 517 | 85 | ||
| FQ214772.1 | - | 3 - 4 | 517 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oaz2
Genomic Map