Assay Details
| Gene Symbol: Igfbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013122.2 | NP_037254.2 | 1 - 2 | 461 | 108 | |
| GenBank mRNA | BC092570.1 | - | 1 - 2 | 461 | 7308779 | 108 |
| J04486.1 | - | 1 - 2 | 108 | |||
| M31672.1 | - | 1 - 2 | 108 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Igfbp2
Genomic Map