Assay Details
| Gene Symbol: Casp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271984.1 | NP_001258913.1 | 3 - 4 | 248 | 74 | |
| NM_031775.3 | NP_113963.2 | 4 - 5 | 403 | 74 | ||
| XM_008761527.1 | XP_008759749.1 | 2 - 3 | 193 | 74 | ||
| XM_008761528.1 | XP_008759750.1 | 2 - 3 | 193 | 74 | ||
| GenBank mRNA | AF025670.1 | - | 4 - 5 | 74 | ||
| BC078785.1 | - | 4 - 5 | 368 | 7123179 | 74 | |
| DY310578.1 | - | 3 - 4 | 235 | 74 | ||
| FM068086.1 | - | 3 - 4 | 248 | 74 | ||
| Gene Symbol: LOC103689977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233278.2 | XP_006233340.1 | 4 - 5 | 428 | 74 | |
| XM_006233279.2 | XP_006233341.1 | 3 - 4 | 250 | 74 | ||
| XM_008761476.1 | XP_008759698.1 | 4 - 5 | 430 | 74 | ||
| XM_008761477.1 | XP_008759699.1 | 3 - 4 | 253 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Casp6
Genomic Map