Assay Details
| Gene Symbol: Rhov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138542.2 | NP_612551.2 | 3 - 4 | 412 | 66 | |
| XM_008762077.1 | XP_008760299.1 | 3 - 4 | 719 | 66 | ||
| GenBank mRNA | AF097887.1 | - | 2 - 3 | 367 | 66 | |
| BC086990.1 | - | 3 - 4 | 412 | 7135643 | 66 | |
More Information
Set Membership: |
|

Rhov
Genomic Map